missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF4 Antibody (ARHGEF-08), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-62217JF549
This item is not returnable.
View return policy
Description
ARHGEF4 Monoclonal specifically detects ARHGEF4 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| ARHGEF4 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ARHGEF4 | |
| Recombinant fragment of human ARHGEF4 (amino acids 143-271) | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-174 recognizes CD11b antigen (Mac-1 alpha), a 165-170 kDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| ARHGEF-08 | |
| Western Blot, Flow Cytometry | |
| APC-stimulated guanine nucleotide exchange factor 1, Asef, ASEF1, ASEFAPC-stimulated guanine nucleotide exchange factor, GEF4, KIAA1112, Rho guanine nucleotide exchange factor (GEF) 4, rho guanine nucleotide exchange factor 4, STM6 | |
| Mouse | |
| Protein A purified | |
| Signal Transduction | |
| 50649 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction