missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557027
This item is not returnable.
View return policy
Description
Immunogen sequence: HSLQANDTFN KQQWLNCIRQ AKETVLCAAG QAGVLDSEGS FLNPTTGSRE LQGETKLEQM DQSDSESDCS M Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.
Rho-like GTPases are involved in a variety of cellular processes, and they are activated by binding GTP and inactivated by conversion of GTP to GDP by their intrinsic GTPase activity. Guanine nucleotide exchange factors (GEFs) accelerate the GTPase activity of Rho GTPases by catalyzing their release of bound GDP. This gene encodes a guanine nucleotide exchange factor, which specifically activates two members of the Rho GTPase family: RHOA and RHOB, both of which have a role in bone cell biology. It has been identified that genetic variation in this gene plays a role in the determination of bone mineral density (BMD), indicating the implication of this gene in postmenopausal osteoporosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ARHGEF3 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF3 | |
| 1200004I24Rik; 59.8 kDA protein; 9830169H03Rik; ARHG3; ARHGEF3; C76747; DKFZP434F2429; Exchange factor found in platelets and leukemic and neuronal tissues; exchange factor found in platelets and leukemic and neuronal tissues, XPLN; GEF3; MGC118905; Rho guanine nucleotide exchange factor (GEF) 3; Rho guanine nucleotide exchange factor 3; RhoGEF protein; STA3; Xpln | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NR81 | |
| ARHGEF3 | |
| Recombinant protein corresponding to Human ARHGEF3. Recombinant protein control fragment (Product #RP-96425). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction