missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF19 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569914
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rat: 100%.
Guanine nucleotide exchange factors (GEFs) such as ARHGEF19 accelerate the GTPase activity of Rho GTPases (see RHOA, MIM 165390).[supplied by OMIM, Dec 2008].
Specifications
| ARHGEF19 | |
| Polyclonal | |
| Unconjugated | |
| ARHGEF19 | |
| 6030432F23; 6430573B13Rik; Arhgef19; Ephexin-2; Rho guanine nucleotide exchange factor (GEF) 19; Rho guanine nucleotide exchange factor 19; weakly similar to Rho GEF; weakly similar to Rho GEF 5; Wgef | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 128272 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IW93 | |
| ARHGEF19 | |
| synthetic peptide directed towards the middle region of human ARHGEF19. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction