Learn More
ARHGEF18 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11243-1-AP-150UL
 View more versions of this product
Product Code. 16878183
  • £349.00 / 150µL
Description
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
ARHGEF18
Polyclonal
Unconjugated
ARHGEF18
ARHGEF18, KIAA0521, p114 Rho GEF, P114 RhoGEF, p114RhoGEF, SA RhoGEF, Septin associated RhoGEF
Rabbit
Antigen Affinity Chromatography
RUO
102098, 23370
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P9R4, Q6ZSZ5
Arhgef18
ARHGEF18 Fusion Protein Ag1749
150 μL
Primary
Human, Mouse