missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF18 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584879
This item is not returnable.
View return policy
Description
Immunogen sequence: VLESELVQRI QTLSQLLLNL QAVIAHQDSY VETQRAAIQE REKQFRLQST RGNLLLEQER QRNFEKQR Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 91%.
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ARHGEF18 | |
| Polyclonal | |
| Unconjugated | |
| Arhgef18 | |
| 114 kDa Rho-specific guanine nucleotide exchange factor; AI467246; Arhgef18; D030053O22Rik; KIAA0521; p114RhoGEF; P114-RhoGEF; p114-Rho-GEF; rho guanine nucleotide exchange factor 18; rho/rac guanine nucleotide exchange factor (GEF) 18; Rho/Rac guanine nucleotide exchange factor 18; Rho-specific guanine nucleotide exchange factor p114; SA-RhoGEF; Septin-associated RhoGEF | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23370 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZSZ5 | |
| Arhgef18 | |
| Recombinant protein corresponding to Human ARHGEF18. Recombinant protein control fragment (Product #RP-105550). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu