missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGEF18 Monoclonal Antibody (GT1972)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527826
This item is not returnable.
View return policy
Description
Positive Control: Neuro2A, GL261, C8D30, NIH-3T3, C2C12, PC-12, Rat2, 293T, A431, HeLa, HepG2 Predicted reactivity: Mouse (89%), Rat (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Rho GTPases are GTP binding proteins that regulate a wide spectrum of cellular functions. These cellular processes include cytoskeletal rearrangements, gene transcription, cell growth and motility. Activation of Rho GTPases is under the direct control of guanine nucleotide exchange factors (GEFs). The protein encoded by this gene is a guanine nucleotide exchange factor and belongs to the Rho GTPase GFE family. Family members share a common feature, a Dbl (DH) homology domain followed by a pleckstrin (PH) homology domain. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
| ARHGEF18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q6P9R4, Q6ZSZ5 | |
| Arhgef18 | |
| Recombinant protein encompassing a sequence within the center region of human ARHGEF18. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| GT1972 | |
| Unconjugated | |
| Arhgef18 | |
| 114 kDa Rho-specific guanine nucleotide exchange factor; AI467246; Arhgef18; D030053O22Rik; KIAA0521; p114RhoGEF; P114-RhoGEF; p114-Rho-GEF; rho guanine nucleotide exchange factor 18; rho/rac guanine nucleotide exchange factor (GEF) 18; Rho/Rac guanine nucleotide exchange factor 18; Rho-specific guanine nucleotide exchange factor p114; SA-RhoGEF; Septin-associated RhoGEF | |
| Mouse | |
| Protein G | |
| RUO | |
| 102098, 23370, 304193 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction