missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGEF11 Recombinant Rabbit Monoclonal Antibody (1I7H7)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA557914

 View more versions of this product

Product Code. 30283352

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: QGVDQSPKPL IIGPEEDYDP GYFNNESDII FQDLEKLKSR PAHLGVFLRY IFSQADPSPL LFYLCAEVYQ QASPKDSRSL GKDIWNIFLE KNAPLRVKIP EMLQAEIDSR LRNSEDARGV LCEAQEAAMP EIQEQIHDYR TKRTLGLGSL YGENDLLDLD GDPLRERQVA EKQLAALGDI LSKYEEDRSA PMDFALNTYM SHAGIRLREA.

ARHGEF11 is a 1527 amino acid protein with tandem dbl homology and pleckstrin homology domains, PDZ domain, two proline rich sequences and a regulatory G protein-signaling (RGS) sequence. It acts as guanine nucleotide exchange factor (GEF) for RhoA GTPase, but not for RAC1 or CDC42 and acts as GTPase-activating protein (GAP) for GNA12 and GNA13. It forms a complex with G proteins and stimulates neurite through retraction Rho-dependent signals. ARHGEF11 increases glutamate transport 4-fold. It interacts with EAAT4 and modulates its uptake activity and perisynaptic distribution at glutamatergic synapses. It is known to induce the reorganization of the actin cytoskeleton and its overexpression induces the formation of membrane ruffling and filopodia. It is ubiquitously expressed, in brain, lung, liver, kidney and skeletal muscle.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ARHGEF11
Recombinant Monoclonal
0.97 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Arhgef11; B930073M02; E130307F09; glutamate transporter EAAT4 associated protein 48; glutamate transporter EAAT4-associated protein 48; Gtrap48; KIAA0380; mKIAA0380; PDZ-RhoGEF; Prg; Rho guanine exchange factor (GEF) 11; Rho guanine nucleotide exchange factor (GEF) 11; rho guanine nucleotide exchange factor 11; RhoA-specific guanine nucleotide exchange factor; RhoGEF; RhoGEF glutamate transport modulator; RhoGEF glutamate transport modulator GTRAP48; RP11-356J7.2
Rabbit
Protein A
RUO
213498
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
1I7H7
Unconjugated
ARHGEF11
ARHGEF11
Recombinant fusion protein containing a sequence corresponding to amino acids 281-490 of human ARHGEF11 (NP_055599.1).
100 μL
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.