missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF11, Mouse anti-Human, Clone: 2F4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009826-M02.100ug
This item is not returnable.
View return policy
Description
Sequence: RSESLKGREEMKRSRKAENVPRSRSDVDMDAAAEATRLHQSASSSTSSLSTRSLENPTPPFTPKMGRRSIESPSLGFCTDTLLPHLLEDDLGQLSDLEPESpecifications
| ARHGEF11 | |
| Monoclonal | |
| Unconjugated | |
| Rho guanine nucleotide exchange factor (GEF) 11 | |
| DKFZp667F1223/GTRAP48/KIAA0380/PDZ-RHOGEF | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2F4 | |
| In 1x PBS, pH 7.4 | |
| BC057394 | |
| ARHGEF11 | |
| ARHGEF11 (AAH57394, 651 a.a. ∼ 750 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9826 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction