missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGEF11 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33497-20ul
This item is not returnable.
View return policy
Description
ARHGEF11 Monoclonal antibody specifically detects ARHGEF11 in Mouse samples. It is validated for ELISA,Western Blot
Specifications
| ARHGEF11 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| DKFZp667F1223, glutamate transporter EAAT4-associated protein 48, GTRAP48, KIAA0380rho guanine nucleotide exchange factor 11, PDZ-RhoGEF, Rho guanine exchange factor (GEF) 11, Rho guanine nucleotide exchange factor (GEF) 11, RhoA-specific guanine nucleotide exchange factor, RhoGEF glutamate transport modulator | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 281-490 of human ARHGEF11 (NP_055599.1).,, Sequence:, QGVDQSPKPLIIGPEEDYDPGYFNNESDIIFQDLEKLKSRPAHLGVFLRYIFSQADPSPLLFYLCAEVYQQASPKDSRSLGKDIWNIFLEKNAPLRVKIPEMLQAEIDSRLRNSEDARGVLCEAQEAAMPEIQEQIHDYRTKRTLGLGSLYGENDLLDLDGDPLRERQVAEKQLAALGDILSKYEEDRSAPMDFALNTYMSHAGIRLREA | |
| 20 μL | |
| Primary | |
| Mouse | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9826 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction