missing translation for 'onlineSavingsMsg'
Learn More

ARHGEF10 Antibody (6G5), Novus Biologicals™

Product Code. 18369448 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369448 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369448 Supplier Novus Biologicals Supplier No. H00009639M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARHGEF10 Monoclonal antibody specifically detects ARHGEF10 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGEF10
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 6G5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_055444
Gene Alias DKFZp686H0726, GEF10, KIAA0294MGC131664, Rho guanine nucleotide exchange factor (GEF) 10, rho guanine nucleotide exchange factor 10
Host Species Mouse
Immunogen ARHGEF10 (NP_055444, 792 a.a. ~ 890 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GKQDKSGRPTFFTAVFNTFTPAIKESWVNSLQMAKLALEEENHMGWFCVEDDGNHIKKEKHPLLVGHMPVMVAKQQEFKIECAAYNPEPYLNNESQPDS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9639
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.