missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGDIG Polyclonal Antibody

Product Code. 16322005
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16322005 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16322005 Supplier Invitrogen™ Supplier No. PA585053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DKEGGPPAVD EVLDEAVPEY RAPGRKSLLE IRQLDPDDRS LAKYKRVLLG PLPPAVDPSL PNV Highest antigen sequence indentity to the following orthologs: Mouse - 70%, Rat - 42%.

The Ras superfamily of small GTP-binding proteins are critical mediators of diverse cell signaling pathways, including those leading to proliferation, cytoskeletal organization and secretion. The counter-conversion of the active GTPbound form of these proteins to their inactive GDP-bound form is influenced by two types of regulatory proteins: those that alter the intrinsic GTPase activity of the GTP-binding proteins and those that alter the rate of GDP/GTP exchange. Guanine nucleotide-releasing factors (GRFs) increase the GDP dissociation rate, while GDP-dissociation inhibitors (GDIs) decrease the dissociation rate. The Rho GDI subfamily is composed of Rho GDIalpha, Ly-GDI (also known as Rho GDIbeta and previously known as GDI/D4) and Rho GDIgamma. The Rho GDI proteins interact with and have varying affinities for several Ras-like GTP binding proteins, including Rho A, Rho B, Rac and Cdc42. Ly-GDI is expressed only in hematopoietic cells, predominantly in B and T lymphocyte cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGDIG
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ARHGDIG
Gene Accession No. Q99819
Gene Alias Arhgdig; Gdi5; guanosine diphosphate (GDP) dissociation inhibitor 5, Rho associated; rho GDI 3; Rho GDIgamma; Rho GDP dissociation inhibitor (GDI) gamma; Rho GDP dissociation inhibitor gamma; rho GDP-dissociation inhibitor 3; RhoGDI gamma; Rho-GDI gamma; Rho-GDI2; Rho-GDI-3; RHOGDI-3; Rho-GDIgamma; RIP2
Gene Symbols ARHGDIG
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ARHGDIG. Recombinant protein control fragment (Product #RP-101622).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 398
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.