missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ D4-GDI Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100123
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARHGDIB.
Acts as a ventralizing factor during embryogenesis. Inhibits axin-mediated JNK activation by binding axin and disrupting axin homodimerization. This in turn antagonizes a Wnt/beta-catenin-independent dorsalization pathway activated by AXIN/JNK-signaling.
Specifications
| D4-GDI | |
| Polyclonal | |
| Unconjugated | |
| ARHGDIB | |
| ARHGDIB; D4; d4 gdi; GDI LYGDI; GDIA2; GDID4; LYGDI; Ly-GDI; RAP1GN1; Rho GDI 2; Rho GDP dissociation inhibitor (GDI) beta; Rho GDP dissociation inhibitor beta; rho GDP-dissociation inhibitor 2; Rho, GDP dissociation inhibitor (GDI) beta; rho-GDI beta; RhoGDI2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11857, 362456, 397 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52566, Q61599 | |
| ARHGDIB | |
| A synthesized peptide derived from human ARHGDIB(Accession P52566), corresponding to amino acid residues E3-L53. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction