missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567065
This item is not returnable.
View return policy
Description
Immunogen sequence: AYKEFDRKDGD LTMWPRLVS Highest antigen sequence identity to the following orthologs - mouse 40%, rat 45%.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
| ARHGAP8 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP8 | |
| 3110043J09Rik; ARHGAP8; BCH domain-containing Cdc42GAP-like protein; BNIP-2 and Cdc42GAP homology domain-containing, proline-rich and Cdc42GAP-like protein subtype-1; BPGAP1; PP610; Rho GTPase activating protein 8; rho GTPase-activating protein 8; Rho-type GTPase-activating protein 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23779 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P85298 | |
| ARHGAP8 | |
| Recombinant Human ARHGAP8. Recombinant protein control fragment (Product #RP-105483). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction