missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £484.00
Specifications
| Antigen | ARHGAP8 |
|---|---|
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18247232
|
Novus Biologicals
NBP2-57680 |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623038
|
Novus Biologicals
NBP2-57680-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP8 Polyclonal specifically detects ARHGAP8 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ARHGAP8 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 23779 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:AYKEFDRKDGDLTMWPRLVS | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| BCH Domain-Containing Cdc42GAP-Like Protein, BNIP-2 And Cdc42GAP Homology Domain-Containing, Proline-Rich And BPGAP1, Cdc42GAP-Like Protein Subtype-1, PP610, Rho GTPase Activating Protein 8, Rho GTPase-Activating Protein 8, Rho-Type GTPase-Activating Protein 8 | |
| ARHGAP8 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title