missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals ARHGAP6 Antibody (6B3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00000395-M01-100ug
This item is not returnable.
View return policy
Description
ARHGAP6 Monoclonal antibody specifically detects ARHGAP6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
Specifications
| ARHGAP6 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| Rho GTPase activating protein 6, rho GTPase-activating protein 6, RHOGAP6, RHOGAPX-1, Rho-type GTPase-activating protein 6, Rho-type GTPase-activating protein RhoGAPX-1 | |
| ARHGAP6 (NP_038286, 531 a.a. ~ 636 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RHADDNISKDGQEVTGNKMTSLNLATIFGPNLLHKQKSSDKEFSVQSSARAEESTAIIAVVQKMIENYEALFMVPPDLQNEVLISLLETDPDVVDYLLRRKASQSS | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA | |
| 6B3 | |
| Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA | |
| NP_038286 | |
| Mouse | |
| IgG purified | |
| RUO | |
| 395 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction