missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ARHGAP6 Antibody (1G5), Novus Biologicals™

Product Code. 18344628 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18344628 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18344628 Supplier Novus Biologicals™ Supplier No. H00000395M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ARHGAP6 Monoclonal antibody specifically detects ARHGAP6 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP6
Applications ELISA, Western Blot, Sandwich ELISA
Classification Monoclonal
Clone 1G5
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_038286
Gene Alias Rho GTPase activating protein 6, rho GTPase-activating protein 6, RHOGAP6, RHOGAPX-1, Rho-type GTPase-activating protein 6, Rho-type GTPase-activating protein RhoGAPX-1
Host Species Mouse
Immunogen ARHGAP6 (NP_038286, 531 a.a. ∽ 636 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RHADDNISKDGQEVTGNKMTSLNLATIFGPNLLHKQKSSDKEFSVQSSARAEESTAIIAVVQKMIENYEALFMVPPDLQNEVLISLLETDPDVVDYLLRRKASQSS
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 395
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.