missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5105721
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARHGAP42.
This gene encodes a Rho GTPase-activating protein (RhoGAP), and member of the GRAF or BAR-PH family of proteins. Expression of this gene is enriched in vascular smooth muscle cells and the encoded protein inhibits RhoA activity to regulate vascular tone and control blood pressure. A mutation in the first intron of this gene modulates its expression and is associated with reduced blood pressure in human patients with borderline hypertension.
Specifications
| ARHGAP42 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP42 | |
| 9030420J04Rik; Arhgap42; BE136619; Graf3; RGD1564081; Rho GTPase activating protein 42; Rho GTPase-activating protein 10-like; Rho GTPase-activating protein 42; rho-type GTPase-activating protein 42; TMEM133; VRho GTPase activating protein 42 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 143872 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6NI28 | |
| ARHGAP42 | |
| A synthesized peptide derived from human ARHGAP42(Accession A6NI28), corresponding to amino acid residues I351-C401. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction