missing translation for 'onlineSavingsMsg'
Learn More

ARHGAP39 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18420557 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420557 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420557 Supplier Novus Biologicals Supplier No. NBP281940AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ARHGAP39 Polyclonal antibody specifically detects ARHGAP39 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP39
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50mM Sodium Borate
Gene Alias CrGAP, CrossGAP Homolog, CrossGAP Homolog (Drosophila), KIAA1688, Rho GTPase activating protein 39, RhoGAP93B Homolog, RhoGAP93B Homolog (Drosophila), Vilse
Host Species Rabbit
Immunogen ARHGAP39 antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human ARHGAP39. The immunogen is located within amino acids 880 - 930 of ARHGAP39. Amino Acid Squence: MGMQRERYPERQLPWVQT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 80728
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.