missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ARHGAP36 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17116873
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17116873 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17116873 Supplier Invitrogen™ Supplier No. PA5107063

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence homology: Cow: 93%; Dog: 93%; Goat: 93%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Pig: 86%; Sheep: 93%.

GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARHGAP36
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene ARHGAP36
Gene Accession No. Q6ZRI8
Gene Alias ARHGAP36; Rho GTPase activating protein 36; rho GTPase-activating protein 36
Gene Symbols ARHGAP36
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RP13-102H20.1 (aa 468-518).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 158763
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.