missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP36 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531619
This item is not returnable.
View return policy
Description
Predicted reactivity: Mouse (91%), Sheep (94%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
| ARHGAP36 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP36 | |
| ARHGAP36; Rho GTPase activating protein 36; rho GTPase-activating protein 36 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158763 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q6ZRI8 | |
| ARHGAP36 | |
| Recombinant protein encompassing a sequence within the center region of human ARHGAP36. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction