missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575802
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ARHGAP30 is GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
| ARHGAP30 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP30 | |
| 6030405P05Rik; ARHGAP30; Gm102; mFLJ00267; RGD1561419; Rho GTPase activating protein 30; rho GTPase-activating protein 30; Rho-type GTPase-activating protein 30 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 226652, 257106, 498282 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q640N3, Q7Z6I6 | |
| ARHGAP30 | |
| Synthetic peptide corresponding to residues in Human ARHGAP30. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction