missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP30 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£139.00 - £299.00
Specifications
| Antigen | ARHGAP30 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18305052
|
Bio-Techne
NBP3-15556-20UL |
20 μg |
£139.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18319933
|
Novus Biologicals
NBP3-15556-100UL |
100 μg |
£299.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP30 Polyclonal antibody specifically detects ARHGAP30 in Human, Mouse samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| ARHGAP30 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Rho GTPase activating protein 30 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 485-585 of human ARHGAP30 (NP_859071.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| GPCR, Signal Transduction | |
| PBS, 50% glycerol, pH7.3 | |
| 257106 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title