missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP29 Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12583-1-AP-150UL
This item is not returnable.
View return policy
Description
The ARHGAP29 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ARHGAP29. This antibody recognizes human antigen. The ARHGAP29 antibody has been validated for the following applications: WB, IHC, ELISA analysis.Specifications
| ARHGAP29 | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9411 | |
| Store at -20°C. Stable for one year after shipment. | |
| Polyclonal Antibody | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| PARG1; RP11-255E17.1 | |
| Recombinant Protein | |
| 150 μL | |
| Primary | |
| Human | |
| 140 kDa | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction