missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £470.00
Specifications
| Antigen | ARHGAP23 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18460941
|
Novus Biologicals
NBP1-93587-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18049466
|
Novus Biologicals
NBP1-93587 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP23 Polyclonal specifically detects ARHGAP23 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ARHGAP23 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Q9P227 | |
| 57636 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:MNLGFGDESPEPEASGRGERLGRKVAPLATTEDSLASIPFIDEPTSPSIDLQAKHVPASAVVSSAMNSAPVLGTSPSS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Rho GTPase activating protein 23, rho-type GTPase-activating protein 23 | |
| ARHGAP23 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title