missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP22, p Ser22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-44072
This item is not returnable.
View return policy
Description
ARHGAP22 Polyclonal specifically detects ARHGAP22 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ARHGAP22 (p Ser22) | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with No Preservative | |
| Rho GTPase activating protein 22, rho GTPase-activating protein 22, RHOGAP2, Rho-type GTPase-activating protein 22 | |
| Rabbit | |
| 77.78 kDa | |
| 0.05 mg | |
| Diabetes Research, Lipid and Metabolism, Phospho Specific | |
| 58504 | |
| Reconstitute in 185μL of water if being used immediately. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1.07 mg/mL | |
| Western Blot 1 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
| NP_722495, Q8BL80 | |
| ARHGAP22 | |
| ARHGAP22 [p Ser22] Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic phospho-peptide corresponding to the region surrounding mouse pS22 region of ARHGAP22. (Uniprot: Q8BL80) | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is specific for phosphorylated ARHGAP22 at Serine 22. It also recognizes the S397->A mutation but not the S22->A mutation. A BLAST analysis was used to suggest cross-reactivity with Mouse, Rat and Human based on 100% sequence homology. Cross-reactivity with ARHGAP22 pS22 from other sources has not been determined. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction