missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP21 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100388
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ARHGAP10.
Functions as a GTPase-activating protein (GAP) for RHOA and CDC42. Downstream partner of ARF1 which may control Golgi apparatus structure and function. Also required for CTNNA1 recruitment to adherens junctions.
Specifications
| ARHGAP21 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP21 | |
| 5530401C11Rik; AA416458; ARHGAP10; ARHGAP21; Kiaa1424; Rho GTPase activating protein 21; rho GTPase-activating protein 10; Rho GTPase-activating protein 21; Rho-GTPase activating protein 10; rho-type GTPase-activating protein 21 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57584, 71435 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5T5U3, Q6DFV3 | |
| ARHGAP21 | |
| A synthesized peptide derived from human ARHGAP21(Accession Q5T5U3), corresponding to amino acid residues A416-V466. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction