missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP20 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104102
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RHG20.
ARHGAP20 is a potent GTPase-activating protein (GAP) and convert Rho-type GTPases into an inactive GDP-bound state. It contains a pleckstrin homology (PH) domain, a Ras-associating domain, a Rho-GAP domain and two Annexin-like (ANXL) repeats. ARHGAP20 is a direct downstream target of Rap1 in the neurite outgrowth while it's over expression leads to inactivation of Rho for promoting the neurite outgrowth in a Rap1-dependent manner. ARHGAP20 is a tumor suppressor gene and is inactivated by deletion in breast cancer and also by chromosomal translocation in B-cell chronic lymphocytic leukemia. Reports suggest that it may take part in rearrangements of the cytoskeleton and cell signaling events that occur during spermatogenesis. It is expressed predominantly in human brain, liver, ovary and spinal cord while low expression is found in lymph nodes and fetal liver.
Specifications
| ARHGAP20 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP20 | |
| 6530403F17Rik; A530023E23Rik; Arhgap20; KIAA1391; mKIAA1391; RA and RhoGAP domain containing protein; RA and RhoGAP domain-containing protein; RARhoGAP; Rho GTPase activating protein 20; rho GTPase activating protein 20 variant 2; rho GTPase-activating protein 20; RhoGTPase activating protein; Rho-type GTPase-activating protein 20 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 244867, 367085, 57569 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q6IFT4, Q6REY9, Q9P2F6 | |
| ARHGAP20 | |
| A synthesized peptide derived from human ARHGAP20(Accession Q9P2F6), corresponding to amino acid residues Q503-E553. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction