missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP19 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12578-100ul
This item is not returnable.
View return policy
Description
ARHGAP19 Polyclonal antibody specifically detects ARHGAP19 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| ARHGAP19 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-5uL/mg of uLysate, Immunohistochemistry-Paraffin 1:100-1:500 | |
| DKFZp313K217, MGC138804, MGC138805, MGC14258, putative RhoGAP protein, Rho GTPase activating protein 19, rho GTPase-activating protein 19, rho-type GTPase-activating protein 19 | |
| Produced in rabbits immunized with E. coli-derived Human ARHGAP19 fragment | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 84986 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction