missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP18 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81704IR
This item is not returnable.
View return policy
Description
ARHGAP18 Polyclonal antibody specifically detects ARHGAP18 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ARHGAP18 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 755 | |
| bA307O14.2, MacGAP, MGC126757, Rho GTPase activating protein 18, rho-type GTPase-activating protein 18 | |
| ARHGAP18 antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human ARHGAP18. The immunogen is located within the last 50 amino acids of ARHGAP18. Amino Acid Squence: DTYMKDLYQLNPNAE | |
| 0.1 mL | |
| Signal Transduction | |
| 93663 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction