missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP17 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559683
This item is not returnable.
View return policy
Description
Immunogen sequence: KQLARLVLDW DSVRARWNQA HKSSGTNFQG LPSKIDTLKE EMDEAGNKVE QCKDQLAADM YNFMAKEGEY GKFFVTLLEA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
RICH1 is a GTPase-activating protein (GAP). GAPs stimulate the intrinsic GTP hydrolysis of small G proteins, such as RHOA (MIM 165390), RAC1 (MIM 602048), and CDC42 (MIM 116952).
Specifications
| ARHGAP17 | |
| Polyclonal | |
| Unconjugated | |
| Arhgap17 | |
| 5730403H17Rik; Arhgap17; MST066; MST110; MSTP038; MSTP066; MSTP110; Nadrin; Nadrin2; neuron-associated developmentally regulated protein; neuron-associated developmentally-regulated protein; neuron-specific GTPase activating protein; neuron-specific GTPase-activating protein; PP367; PP4534; Rho GTPase activating protein 17; rho GTPase-activating protein 17; RhoGAP interacting with CIP4 homologs 1; RhoGAP interacting with CIP4 homologs protein 1; Rho-type GTPase-activating protein 17; Rich1; RICH-1; RICH1B; WBP15; WW domain-binding protein 15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55114, 70497 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q3UIA2, Q68EM7 | |
| Arhgap17 | |
| Recombinant protein corresponding to Human ARHGAP17. Recombinant protein control fragment (Product #RP-98644). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction