missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP17 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97377PCP
This item is not returnable.
View return policy
Description
ARHGAP17 Polyclonal antibody specifically detects ARHGAP17 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| ARHGAP17 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| DKFZp564A1363, FLJ10308, FLJ13219, FLJ37567, FLJ43368, MGC87805, MST110, MSTP038, MSTP066, MSTP110, NADRIN, neuron-associated developmentally regulated protein, PP367, PP4534, Rho GTPase activating protein 17, rho GTPase-activating protein 17, RhoGAP interacting with CIP4 homologs 1, RhoGAP interacting with CIP4 homologs protein 1, Rho-type GTPase-activating protein 17, RICH-1, RICH1B, RICH1MST066, WBP15 | |
| Produced in rabbits immunized with E. coli-derived Human ARHGAP17 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55114 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction