missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHGAP12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104101
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total RHG12.
GTPase activator for the Rho-type GTPases by converting them to an inactive GDP-bound state.
Specifications
| ARHGAP12 | |
| Polyclonal | |
| Unconjugated | |
| ARHGAP12 | |
| 2810011M08Rik; ARHGAP12; FLJ10971; FLJ20737; FLJ21785; Rho GTPase activating protein 12; rho GTPase-activating protein 12; Rho-type GTPase-activating protein 12 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 307016, 75415, 94134 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8C0D4, Q8IWW6 | |
| ARHGAP12 | |
| A synthesized peptide derived from human ARHGAP12(Accession Q8IWW6), corresponding to amino acid residues S338-L388. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction