missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£222.00 - £451.00
Specifications
| Antigen | ARHGAP12 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18482622
|
Novus Biologicals
NBP1-91678-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18731724
|
Novus Biologicals
NBP1-91678 |
0.1 mL |
£451.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP12 Polyclonal specifically detects ARHGAP12 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ARHGAP12 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q8IWW6 | |
| 94134 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RGTQERTWKPPRWTRDASISKGDFQNPGDQELLSSEENYYSTSYSQSDSQCGSPPRGWSEELDERGHTLYTSDYTNEKWLKHVDDQGRQYYYSADGSRSEWELPKYNASSQQQREIIKSRSLDRRLQEPIVLTKWRHSTIVLDTNDKESPTASKPCFPENESSPSSP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 96 kDa |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Rho GTPase activating protein 12 | |
| ARHGAP12 | |
| IgG | |
| Affinity Purified | |
| Specificity of human ARHGAP12 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title