missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP11B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£174.00 - £255.00
Specifications
| Antigen | ARHGAP11B |
|---|---|
| Dilution | Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18642078
|
Novus Biologicals
NBP3-06142-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18625067
|
Novus Biologicals
NBP3-06142-50ul |
50 μg |
£174.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ARHGAP11B Polyclonal antibody specifically detects ARHGAP11B in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| ARHGAP11B | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 89839 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry-Paraffin 1:200-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| B'-T, FAM7B1, Family With Sequence Similarity 7, Member B1, GAP (1-8), Rho GTPase Activating Protein 11B, Rho GTPase-Activating Protein 11B, Rho-Type GTPase-Activating Protein 11B | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ARHGAP11B. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title