missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARHGAP11B Polyclonal antibody specifically detects ARHGAP11B in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | ARHGAP11B |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | B'-T, FAM7B1, Family With Sequence Similarity 7, Member B1, GAP (1-8), Rho GTPase Activating Protein 11B, Rho GTPase-Activating Protein 11B, Rho-Type GTPase-Activating Protein 11B |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human ARHGAP11B. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?