missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARHGAP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24707
This item is not returnable.
View return policy
Description
ARHGAP1 Polyclonal specifically detects ARHGAP1 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications
| ARHGAP1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.2% Gelatin with 0.05% Sodium Azide | |
| CDC42 GTPase-activating protein, CDC42GAPp50-RhoGAP, GTPase-activating protein rhoOGAP, p50rhoGAP, Rho GTPase activating protein 1, rho GTPase-activating protein 1, RHOGAP, RHOGAP1Cdc42GAP, Rho-related small GTPase protein activator, Rho-type GTPase-activating protein 1 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| 392 | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| Purified |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| Q07960 | |
| ARHGAP1 | |
| A portion of amino acids 1-50 of human RhoGAP1 was used as the immunogen | |
| 0.1 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse and rat (each having multiple isoforms), 94% homologous in bovine, 88% homologous in chicken, 76% homologous in Xenopus, and 70% homologous in zebrafish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction