missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHE Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563863
This item is not returnable.
View return policy
Description
Immunogen sequence: SHVATLACVN KTNKNVKRNK SQRATKRISH MPSRPELSAV ATDLRKDKAK SCTVL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras, Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex; GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins; and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form.
Specifications
| ARHE | |
| Polyclonal | |
| Unconjugated | |
| RND3 | |
| 2610017M01Rik; AI661404; Arhe; memB; MemB protein; Protein MemB; ras homolog gene family, member E; Rho family GTPase 3; Rho related protein Rnd3/Rho8; rho8; RHOE; rho-related GTP-binding protein Rho8; Rho-related GTP-binding protein RhoE; Rnd3; small GTP binding protein Rho8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 390 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P61587 | |
| RND3 | |
| Recombinant protein corresponding to Human ARHE. Recombinant protein control fragment (Product #RP-103783). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction