missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARHE Monoclonal Antibody (5C7E8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531896
This item is not returnable.
View return policy
Description
MA5-31896 has been tested in indirect ELISA.
Members of the Rho family of Ras-related GTPases, such as ARHE, regulate the organization of the actin cytoskeleton in response to extracellular growth factors. Like Ras, Rho family members appear to cycle between an inactive GDP-bound form and an active GTP-bound form. Three major regulators of Rho activity have been identified: RhoGDIs, which interact with the GDP-bound Rho proteins to keep them in a resting complex; GEFs, which promote GDP/GTP exchange leading to activation of Rho proteins; and GAPs, which stimulate GTP hydrolysis and return the activated Rho protein to its inactive form.
Specifications
| ARHE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| P61587 | |
| RND3 | |
| Purified recombinant fragment of human RND3 (AA: 104-241) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5C7E8 | |
| Unconjugated | |
| RND3 | |
| 2610017M01Rik; AI661404; Arhe; memB; MemB protein; Protein MemB; ras homolog gene family, member E; Rho family GTPase 3; Rho related protein Rnd3/Rho8; rho8; RHOE; rho-related GTP-binding protein Rho8; Rho-related GTP-binding protein RhoE; Rnd3; small GTP binding protein Rho8 | |
| Mouse | |
| Protein G | |
| RUO | |
| 390 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction