Learn More
Invitrogen™ LDLRAP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109904
Description
Immunogen sequence: EQPKGEELSA AAIKRIVATA KASGKKLQKV TLKVSPRGII LTDNLTNQLI ENVSIYRISY CTADKMHDKV FAYIAQSQHN QSLECHAFLC TKRKMAQAVT LTVAQAFKVA FEFWQVSKEE KEKRDKASQE GGDVLGARQD CTPSLKSLVA TGNLLDLEET AKAPLSTVSA NTTNMDEVPR PQALSGSSVV WELDDGLDEA FSRLAQSRTN PQVLDTGLTA QDMHYAQCLS PVDWDKPDSS GTEQDDLFSF.
The protein encoded by this gene is a cytosolic protein which contains a phosphotyrosine binding (PTD) domain. The PTD domain has been found to interact with the cytoplasmic tail of the LDL receptor. Mutations in this gene lead to LDL receptor malfunction and cause the disorder autosomal recessive hypercholesterolaemia.
Specifications
| LDLRAP1 | |
| Polyclonal | |
| Unconjugated | |
| LDLRAP1 | |
| AA691260; Arh; Arh1; ARH2; Autosomal recessive hypercholesterolemia protein; autosomal recessive hypercholesterolemia protein homolog; DKFZp586D0624; FHCB1; FHCB2; LDL receptor adaptor protein; LDLRAP1; Low density lipoprotein receptor adapter protein 1; low density lipoprotein receptor adaptor protein 1; MGC34705; RGD1563417; RP11-70P17.2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 100017, 26119, 500564 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.22 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| D3ZAR1, Q5SW96, Q8C142 | |
| LDLRAP1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 59-308 of human LDLRAP1 (NP_056442.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.