Learn More
ARH Mouse anti-Human, Unconjugated, Clone: 1D1B11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66932-2-IG-150UL
This item is not returnable.
View return policy
Description
The ARH antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ARH. This antibody recognizes human, rat antigen. The ARH antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ARH | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat |
| Western Blot, ELISA | |
| 1D1B11 | |
| Unconjugated | |
| 150 μL | |
| 26119 | |
| IgG2b |