missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ARH Monoclonal antibody specifically detects ARH in Human, Mouse, Rat, Canine samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ARH |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI7A5 |
| Conjugate | PE |
| Dilution | Western Blot |
| Formulation | PBS |
| Gene Alias | ARH1, ARH2, ARHAutosomal recessive hypercholesterolemia protein, DKFZp586D0624, FHCB1ARH1, FHCB2LDL receptor adaptor protein, low density lipoprotein receptor adapter protein 1, low density lipoprotein receptor adaptor protein 1, MGC34705 |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human LDLRAP1(NP_056442) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?