missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Argonaute 4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA567021

Product Code. 15979694

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: VQWVSVVSLQL LLEALAGHLN EVPDDSVQAL DVITRH Highest antigen sequence identity to the following orthologs - mouse 100%, rat 57%.

This gene encodes a member of the Argonaute family of proteins which play a role in RNA interference. The encoded protein is highly basic containing PAZ and PIWI domains, and it may play a role in short-interfering-RNA-mediated gene silencing.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Argonaute 4
Polyclonal
Unconjugated
AGO4
5730550L01Rik; AGO4; AI481660; argonaute 4; argonaute 4, RISC catalytic component; argonaute RISC catalytic component 4; argonaute RISC catalytic subunit 4; argonaute RISC component 4; argonaute4; eIF2C 4; eIF-2C 4; Eif2c4; Eukaryotic translation initiation factor 2C 4; eukaryotic translation initiation factor 2C, 4; hAgo4; KIAA1567; LOW QUALITY PROTEIN: protein argonaute-4; mAgo4; Piwi/Argonaute family protein meIF2C4; Protein argonaute-4
Rabbit
Antigen Affinity Chromatography
RUO
192670
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9HCK5
AGO4
Recombinant Human Argonaute 4. Recombinant protein control fragment (Product #RP-105460).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.