Learn More
Invitrogen™ Argonaute 3 Recombinant Superclonal™ Antibody
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711756
Description
This antibody is predicted to react with Monkey, Cat, Sheep, and Bovine. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
AGO3 (protein argonaute-3) is required to RNA-mediated gene silencing (RNAi). It binds to short RNAs such as microRNAs (miRNAs) and represses the translation of mRNAs which are complementary to them. AGO3 is proposed to be involved in stabilization of small RNA derivates (riRNA) derived from processed RNA polymerase III-transcribed Alu repeats containing a DR2 retinoic acid response element (RARE) in stem cells and in the subsequent riRNA-dependent degradation of a subset of RNA polymerase II-transcribed coding mRNAs by recruiting a mRNA decapping complex involving EDC4.
Specifications
| Argonaute 3 | |
| Recombinant Superclonal | |
| Unconjugated | |
| AGO3 | |
| AGO3; argonaute 3; argonaute 3, RISC catalytic component; argonaute RISC catalytic component 3; argonaute RISC catalytic subunit 3; argonaute3; AW048688; C130014L07Rik; eIF2C 3; eIF-2C 3; EIF2C1; EIF2C3; Eukaryotic translation initiation factor 2C 3; eukaryotic translation initiation factor 2C, 1; eukaryotic translation initiation factor 2C, 3; hAgo3; mAgo3; Piwi/Argonaute family protein meIF2C3; Protein argonaute-3 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 192669 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9H9G7 | |
| AGO3 | |
| Peptides corresponding to human AGO3 [1) aa10-aa29 2) aa385-aa404]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.