missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argonaute 3 Antibody (PCRP-AGO3-1C5), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14202CL1
This item is not returnable.
View return policy
Description
Argonaute 3 Monoclonal antibody specifically detects Argonaute 3 in Human samples. It is validated for Flow Cytometry,ELISA,Immunoprecipitation,Protein Array,Immunocytochemistry/Immunofluorescence
Specifications
| Argonaute 3 | |
| Monoclonal | |
| CoraFluor 1 | |
| AGO3argonaute 3, argonaute3, eIF2C 3, eIF-2C 3, Eukaryotic translation initiation factor 2C 3, eukaryotic translation initiation factor 2C, 3, FLJ12765, hAgo3, MGC86946, protein argonaute-3 | |
| Recombinant full-length human Argonaute 3 protein (Uniprot: Q9H9G7) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Protein Array, Immunocytochemistry/Immunofluorescence | |
| PCRP-AGO3-1C5 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 192669 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction