missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AGO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550654
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total AGO1 protein.
AGO1 is a member of the Argonaute family of proteins which play a role in RNA interference. The protein is highly basic, and contains a PAZ domain and a PIWI domain. It may interact with dicer1 and play a role in short-interfering-RNA-mediated gene silencing. AGO1 binds to miRNAs or siRNAs and represses the translation of mRNAs.
Specifications
| AGO1 | |
| Polyclonal | |
| Unconjugated | |
| Ago1 | |
| AGO1; argonaute 1; argonaute 1, RISC catalytic component; argonaute RISC catalytic component 1; argonaute RISC catalytic subunit 1; argonaute1; EIF2C; eIF2C 1; eIF-2C 1; EIF2C1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; GERP95; Golgi Endoplasmic Reticulum protein 95 kDa; hAgo1; mAgo1; Piwi/Argonaute family protein meIF2C1; Protein argonaute-1; putative RNA-binding protein Q99; Q99 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 236511, 26523 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 2.6 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q8CJG1, Q9UL18 | |
| Ago1 | |
| Synthetic peptide corresponding to a region derived from internal residues of human argonaute RISC catalytic component 1. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction