missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AGO1 Monoclonal Antibody (OTI4G1)

AGO1 Monoclonal Antibody (OTI4G1)
Brand: Invitrogen MA527411
This item is not returnable.
View return policy
Description
AGO1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
| AGO1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UL18 | |
| Ago1 | |
| Human recombinant protein fragment corresponding to amino acids 226-490 of EIF2C1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G1 | |
| Unconjugated | |
| Ago1 | |
| AGO1; argonaute 1; argonaute 1, RISC catalytic component; argonaute RISC catalytic component 1; argonaute RISC catalytic subunit 1; argonaute1; EIF2C; eIF2C 1; eIF-2C 1; EIF2C1; Eukaryotic translation initiation factor 2C 1; eukaryotic translation initiation factor 2C, 1; GERP95; Golgi Endoplasmic Reticulum protein 95 kDa; hAgo1; mAgo1; Piwi/Argonaute family protein meIF2C1; Protein argonaute-1; putative RNA-binding protein Q99; Q99 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 26523, 313594 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction