missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arginyl tRNA synthetase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | Arginyl tRNA synthetase |
|---|---|
| Dilution | Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231428
|
Novus Biologicals
NBP3-38241-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227484
|
Novus Biologicals
NBP3-38241-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Arginyl tRNA synthetase Polyclonal antibody specifically detects Arginyl tRNA synthetase in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ImmunofluorescenceSpecifications
| Arginyl tRNA synthetase | |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| arginine tRNA ligase 1, cytoplasmic, Arginine--tRNA ligase, arginyl-tRNA synthetase, arginyl-tRNA synthetase, cytoplasmic, ArgRS, DALRD1, EC 6.1.1.19, MGC8641 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Arginyl tRNA synthetase (NP_002878.2).,, Sequence:, MDVLVSECSARLLQQEEEIKSLTAEIDRLKNCGCLGASPNLEQLQEENLKLKYRLNILRKSLQAERNKPTKNMINIISRLQEVFGHAIKAAYPDLENPPLLVTPSQQAKFGDYQCNSAMGISQMLKTKEQKVNPREIAENITKHLPDNECIEKVEIAGPGFINVHLRKDFVSEQLTSLLVNGVQLPALGENKKVIVDFSSPNIAKEMHVGHLRSTIIGESISRLFEFAGYDVLRLNHVGDWGTQFGMLIAHLQDKFPDYLTVSPPIGDLQVFYKESKKRFDTEEEFKKRAYQCVVLLQGK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:1000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:10 - 1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 5917 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title