missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Synthase Antibody (OTI1B10), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70217H
This item is not returnable.
View return policy
Description
Argininosuccinate Synthase Monoclonal specifically detects Argininosuccinate Synthase in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Argininosuccinate Synthase | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| ASS1 | |
| Synthetic peptide corresponding to residues near C-terminus of human Argininosuccinate Synthase (NP_000041). | |
| 0.1 mL | |
| 445.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1B10 | |
| Western Blot | |
| argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction