missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Argininosuccinate Synthase Antibody (1G11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-59443CL1
This item is not returnable.
View return policy
Description
Argininosuccinate Synthase Monoclonal antibody specifically detects Argininosuccinate Synthase in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| Argininosuccinate Synthase | |
| Monoclonal | |
| CoraFluor 1 | |
| argininosuccinate synthase, argininosuccinate synthase 1, argininosuccinate synthetase, argininosuccinate synthetase 1, ASSCTLN1, citrulline-aspartate ligase, Citrulline--aspartate ligase, EC 6.3.4.5 | |
| Recombinant human Argininosuccinate Synthase (1-412aa) purified from E. coli | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 445 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 1G11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction