missing translation for 'onlineSavingsMsg'
Learn More

Arginine, Mikromol™

Product Code. 17919684
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17919684 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919684 Supplier Mikromol Supplier No. MM0568.00
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Arginine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Arginine
CAS 74-79-3
Analyte or Component Names L-Arginine
Shipping Condition Room Temperature
Molecular Formula C6 H14 N4 O2
Certifications/Compliance ISO 17034
Synonym Arginine, L-Arginine (9CI, ACI), Arginine, L- (8CI), (S)-2-Amino-5-[(aminoiminomethyl)amino]pentanoic acid, Arginine, L-(+)-Arginine, L-Arg, 5-[(Aminoiminomethyl)amino]-L-norvaline, N5-(Aminoiminomethyl)-L-ornithine, NSC 206269, L-alpha-Amino-delta-guanidinovaleric acid, L-α-Amino-Δ-guanidinovaleric acid, (2S)-2-Amino-5-(diaminomethylideneamino)pentanoic acid, 117: PN: WO2021061872 SEQID: 117 claimed protein, 381: PN: WO2022056313 SEQID: 385 claimed protein, 42: PN: KR20200014684 SEQID: 105 claimed sequence, 42: PN: WO2020027640 SEQID: 105 claimed protein, 87: PN: WO2022010798 SEQID: 89 claimed protein, L-Norvaline, 5-[(aminoiminomethyl)amino]-, L-Ornithine, N5-(aminoiminomethyl)-, A 0526, L-arginine C Grade, Pentanoic acid, 2-amino-5-[(aminoiminomethyl)amino]-, (S)
InChI Formula InChI=1S/C6H14N4O2/c7-4(5(11)12)2-1-3-10-6(8)9/h4H,1-3,7H2,(H,11,12)(H4,8,9,10)/t4-/m0/s1
SMILES N[C@@H](CCCNC(=N)N)C(=O)O
IUPAC Name (2S)-2-amino-5-carbamimidamidopentanoic acid
Molecular Weight (g/mol) 174.2
Quantity 500 mg
Formula Weight 174.1117 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.